Tetracaine 98%

Supplier: Thermo Scientific
Danger

Synonyms: 4-(Butylamino)benzoic Acid 2-(Dimethylamino)ethyl Ester, 2-(Dimethylamino)ethyl 4-(Butylamino)benzoate

460820050 460820250 460821000
ACRO460820050EA 50.7 EUR
ACRO460820050 ACRO460820250 ACRO460821000
Tetracaine 98%
Tetracaine
Formula: C₁₅H₂₄N₂O₂
MW: 264,37 g/mol
Melting Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00053787
CAS Number: 94-24-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 41 to 46 °C
Titration with HClO4 97.5 to 102 % (on dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR