Lamotrigine 98%

Supplier: Thermo Scientific
Danger

Synonyms: 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine

460850010 460850050
ACRO460850010EA 67.8 EUR
ACRO460850010 ACRO460850050
Lamotrigine 98%
Lamotrigine
Formula: C₉H₇Cl₂N₅
MW: 256,09 g/mol
Melting Pt: 218 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00865333
CAS Number: 84057-84-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 216 to 220 °C
Loss on drying ≤0.5 % (105 °C, 3 h)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR