Benzbromarone 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(3,5-Dibromo-4-hydroxyphenyl)(2-ethylbenzofuran-3-yl)methanone, 3-(3,5-Dibromo-4-hydroxybenzoyl)-2-ethylbenzofuran
460900010
460900050
ACRO460900010EA
41
EUR
ACRO460900010
ACRO460900050
Benzbromarone 98%
Benzbromarone
Formula:
C₁₇H₁₂Br₂O₃ MW: 424,09 g/mol Melting Pt: 152 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00078962 CAS Number: 3562-84-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 150 to 154 °C |
Loss on drying | ≤0.5 % |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...