Gliclazide
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
N-((Hexahydrocyclopenta[c]pyrrol-2(1H)-yl)carbamoyl)-4-methylbenzenesulphonamide, 1-(Hexahydrocyclopenta[c]pyrrol-2(1H)-yl)-3-[(4-methylphenyl)sulphonyl]urea, 1-[3-Azabicyclo[3.3.0]oct-3-yl]-3-p-toluenesulphonylurea
461070010
461070050
ACRO461070010EA
51
EUR
ACRO461070010
ACRO461070050
Gliclazide
Gliclazide
Formula:
C₁₅H₂₁N₃O₃S MW: 323,41 g/mol Melting Pt: 163…169 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00409893 CAS Number: 21187-98-4 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 163 to 169 °C |
Loss on drying | ≤0.5 % (1 g, 105 °C) |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...