Febuxostat 98%
Supplier: Thermo Fisher Scientific
Synonyms:
2-(3-Cyano-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylic acid
461580010
461580050
ACRO461580010EA
152
EUR
ACRO461580010
ACRO461580050
Febuxostat 98%
Febuxostat
Formula:
C₁₆H₁₆N₂O₃S Melting Pt: 209 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00871598 CAS Number: 144060-53-7 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 207 to 211 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...