Pharmacy/vaccine refrigerators
Supplier: Labcold
RLDF18041 RLDF0110A RLDF1010A RLDG0510A RLDG1010A RLDF0210A RLDG0110A RLDG0210A
471-1102EA 4600 EUR
471-1102 471-0918 471-0920 471-0921 471-0922 471-1100 BORORLDG0110A BORORLDG0210A
Pharmacy/vaccine refrigerators
Refrigerators
Pharmacy refrigerators designed for dispensary, surgery, ward and general use for the storage of drugs and vaccines under controlled conditions in line with the 'Code of Ethics and Standards' of the Royal Pharmaceutical Society of Great Britain. Where necessary, to achieve the required temperature control, circulating fans are fitted. All models feature automatic temperature data logging for up to 13 months, remote alarm contacts and alarm battery back-up.
  • Digital temperature display with max./min. record
  • Lockable door
  • High/low temperature alarm, door open alarm
  • Powered by the unique IntelliCold™ controller for superior temperature uniformity and energy consumption, plus extra functions designed to store temperature sensitive pharmaceuticals in optimum conditions and assist busy professionals with regulatory requirements
  • CFC, HCFC and 100% ammonia free
Temperature: 2 to +8 °C
Conforms to RPS guidelines and to Department of Health "Green Book" requirements.
Certifications: Conforms to RPS guidelines and to Department of Health "Green Book" requirements.