Cesium ≥99.8% (metals basis)
Supplier: Thermo Fisher Scientific
|
Danger
|
43005.03
43005.06
43005.18
43005.03EA
164
EUR
43005.03
43005.06
43005.18
Cesium ≥99.8% (metals basis)
Cesium
Purity: 99.8%
CAS No.: 7440-46-2
Physical Form: Ampouled
Odour: Odourless
UN No.: 1407
MDL No.: MFCD00134037
Formula:
Cs Boiling Pt: 705 °C (1013 hPa) Melting Pt: 28,5 °C Density: 1,873 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134037 CAS Number: 7440-46-2 EINECS: 231-155-4 UN: 1407 ADR: 4.3,I Merck Index: 12,02051 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...