Aluminium ≥99.9995% (metals basis), foil 50x125 mm (2.0x4.9 in), Thickness: 0.13 mm (0.005 in)

Supplier: Thermo Fisher Scientific

00004.KF 00004.KI
00004.KFEA 94.6 EUR
00004.KF 00004.KI
Aluminium ≥99.9995% (metals basis), foil 50x125 mm (2.0x4.9 in), Thickness: 0.13 mm (0.005 in)
Aluminium

Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈2.2g/50x125mm Odour: Odourless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2,7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR