Indium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
11383.FF
11383.FI
11383.GH
11383.FFEA
181
EUR
11383.FF
11383.FI
11383.GH
Indium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
Indium
Purity: 99.998%
CAS No.: 7440-74-6
Physical Form: ≈4.6g/25x25mm
Odour: Odourless
MDL No.: MFCD00134048
Formula:
In Boiling Pt: 2000 °C (1013 hPa) Melting Pt: 156 °C Density: 7,3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...