Germanium 99.999% (trace metals basis), powder
Supplier: Thermo Scientific
|
Warning
|
193110100
193110500
ACRO193110100EA
242
EUR
ACRO193110100
ACRO193110500
Germanium 99.999% (trace metals basis), powder
Germanium
Formula:
Ge MW: 72,61 g/mol Boiling Pt: 2830 °C (1013 hPa) Melting Pt: 937 °C Density: 5,35 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00085310 CAS Number: 7440-56-4 EINECS: 231-164-3 UN: 3089 ADR: 4.1,III Merck Index: 12,04415 |
Specification Test Results
Appearance | Grey to black Glistening crystalline powder |
Total trace metal impurities | ≤0.001 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...