Germanium ≥99.999% (metals basis), pieces ≤ 3.2 mm (0.125 in)
Supplier: Thermo Scientific
43986.09
43986.18
43986.09EA
241
EUR
43986.09
43986.18
Germanium ≥99.999% (metals basis), pieces ≤ 3.2 mm (0.125 in)
Germanium
Purity: 99.999%
CAS No.: 7440-56-4
Odour: Odourless
MDL No.: MFCD00085310
Formula:
Ge MW: 72,61 g/mol Boiling Pt: 2830 °C (1013 hPa) Melting Pt: 937,2 °C Density: 5,323 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085310 CAS Number: 7440-56-4 EINECS: 231-164-3 Merck Index: 12,04415 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...