Silicon ≥99.999% (metals basis), crystalline powder -325 mesh
Supplier: Thermo Fisher Scientific
|
Warning
|
35662.09
35662.18
35662.09EA
107
EUR
35662.09
35662.18
Silicon ≥99.999% (metals basis), crystalline powder -325 mesh
Silicon
Purity: 99.999%
CAS No.: 7440-21-3
Appearance: White
Odour: Odourless
UN No.: 1346
MDL No.: MFCD00085311
Formula:
Si Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS Number: 7440-21-3 EINECS: 231-130-8 UN: 1346 ADR: 4.1,III Merck Index: 12,08635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...