Tin ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
11458.FI
11458.GH
11458.FIEA
125
EUR
11458.FI
11458.GH
Tin ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Tin
Purity: 99.9985%
CAS No.: 7440-31-5
Physical Form: ≈9.1g/50x50mm
Odour: Odourless
MDL No.: MFCD00133862
Formula:
Sn MW: 118.71 g/mol Boiling Pt: 2270 Melting Pt: 232 Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...