Dysprosium ≥99.8% (REO, rare earth oxide basis), ingot
Supplier: Thermo Scientific
00102.14
00102.22
00102.14EA
145
EUR
00102.14
00102.22
Dysprosium ≥99.8% (REO, rare earth oxide basis), ingot
Dysprosium
Purity: 99.8%
CAS No.: 7429-91-6
Physical Form: Packaged in air stable poly bottle
Odour: Odourless
MDL No.: MFCD00010982
Formula:
Dy MW: 162,5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010982 CAS Number: 7429-91-6 EINECS: 231-073-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...