Dysprosium ≥99.8% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)

Supplier: Thermo Fisher Scientific
Danger


Packaged under argon.

00592.FF 00592.FI
00592.FFEA 181 EUR
00592.FF 00592.FI
Dysprosium ≥99.8% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)
Dysprosium

Purity: 99.8% CAS No.: 7429-91-6 Physical Form: ≈3.3g/25x25mm, Packaged under argon Appearance: Gray Odour: Odourless UN No.: 3178 MDL No.: MFCD00010982

Formula: Dy
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9
UN: 3178
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR