Dysprosium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Supplier: Thermo Scientific
|
Danger
|
Packaged under argon.
12257.06
12257.14
12257.22
12257.06EA
127
EUR
12257.06
12257.14
12257.22
Dysprosium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Dysprosium
Purity: 99.9%
CAS No.: 7429-91-6
Physical Form: Packaged under argon
Appearance: Silver
Odour: Odourless
UN No.: 3178
MDL No.: MFCD00010982
Formula:
Dy MW: 162,5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010982 CAS Number: 7429-91-6 EINECS: 231-073-9 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...