Dysprosium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Scientific
|
Danger
|
Packaged under argon.
12382.FF
12382.FI
12382.FFEA
304
EUR
12382.FF
12382.FI
Dysprosium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Dysprosium
Purity: 99.9%
CAS No.: 7429-91-6
Physical Form: ≈0.53g/25x25mm, Packaged under argon
Appearance: Gray
Odour: Odourless
UN No.: 3178
MDL No.: MFCD00010982
Formula:
Dy MW: 162,5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010982 CAS Number: 7429-91-6 EINECS: 231-073-9 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...