Dysprosium ≥99.7% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
40285.DH
40285.DJ
40285.DM
40285.HF
40285.DHEA
314
EUR
40285.DH
40285.DJ
40285.DM
40285.HF
Dysprosium ≥99.7% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Dysprosium
Purity: 99.7%
CAS No.: 7429-91-6
Molecular Formula: Packaged in mylar under argon
Physical Form: ≈0.27g/mm, Cast rod, diameter is approximate
Odour: Odourless
Formula:
Dy MW: 162.5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010982 CAS Number: 7429-91-6 EINECS: 231-073-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...