Erbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)

Supplier: Thermo Fisher Scientific

00593.FF 00593.FI
00593.FFEA 337 EUR
00593.FF 00593.FI
Erbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)
Erbium

Purity: 99.9% CAS No.: 7440-52-0 Physical Form: ≈3.5g/25x25mm Odour: Odourless MDL No.: MFCD00010987

Formula: Er
MW: 167.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR