Lutetium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Fisher Scientific
Danger

00599.FF 00599.FI
00599.FFEA 0 EUR
00599.FF 00599.FI
Lutetium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.127 mm (0.005 in)
Lutetium

Purity: 99.9% CAS No.: 7439-94-3 Physical Form: ≈0.78g/25x25mm, Packaged in mylar under argon Appearance: Silver-grey Odour: Odourless UN No.: 3178 MDL No.: MFCD00011098

Formula: Lu
MW: 174.97 g/mol
Boiling Pt: 3000
Melting Pt: 1651…1675 °C
Density: 9.849 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011098
CAS Number: 7439-94-3
EINECS: 231-103-0
UN: 3178
ADR: 4.1 III
Merck Index: 12,05642

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR