Neodymium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)
Supplier: Thermo Scientific
10248.FF
10248.FI
10248.FFEA
230
EUR
10248.FF
10248.FI
Neodymium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)
Neodymium
Purity: 99.9%
CAS No.: 7440-00-8
Physical Form: ≈4.4g/25x25mm, Packaged in mylar under argon
Odour: Odourless
MDL No.: MFCD00011130
Formula:
Nd MW: 144,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011130 CAS Number: 7440-00-8 EINECS: 231-109-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...