Terbium ≥99.9% (REO, rare earth oxide basis), pieces

Supplier: Thermo Scientific
Danger


Packaged under argon.

00324.03 00324.06
00324.03EA 103 EUR
00324.03 00324.06
Terbium ≥99.9% (REO, rare earth oxide basis), pieces
Terbium

Purity: 99.9% CAS No.: 7440-27-9 Physical Form: Packaged under argon Appearance: Silver-grey Odour: Odourless UN No.: 3178 MDL No.: MFCD00011256

Formula: Tb
MW: 158,93 g/mol
Boiling Pt: 3041 °C (1013 hPa)
Melting Pt: 1356 °C
Density: 8,27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011256
CAS Number: 7440-27-9
EINECS: 231-137-6
ADR: 4.1,II
Merck Index: 12,09300

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR