Terbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Scientific
12429.FF
12429.FI
12429.FFEA
277
EUR
12429.FF
12429.FI
Terbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Terbium
Purity: 99.9%
CAS No.: 7440-27-9
Physical Form: ≈1.29g/25x25mm, Packaged in mylar under argon
MDL No.: MFCD00011256
Formula:
Tb MW: 158,93 g/mol Boiling Pt: 3041 °C (1013 hPa) Melting Pt: 1356 °C Density: 8,27 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011256 CAS Number: 7440-27-9 EINECS: 231-137-6 Merck Index: 12,09300 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...