Silver 99.9%, foil 25x25 mm, Thickness: 1.0 mm , 1ea = 6.6gr
Supplier: Thermo Scientific
317750010
317750040
ACRO317750010EA
86.7
EUR
ACRO317750010
ACRO317750040
Silver 99.9%, foil 25x25 mm, Thickness: 1.0 mm , 1ea = 6.6gr
Silver
Formula:
Ag MW: 107,87 g/mol Boiling Pt: 2212 °C (1013 hPa) Melting Pt: 961 °C Density: 10,5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 Merck Index: 12,08647 |
Specification Test Results
Appearance | Silver white Foil |
Total trace metal impurities | ≤0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...