Mercury
Supplier: Roth Carl
|
Danger
|
8530.2
ROTH8530.2EA
349
EUR
ROTH8530.2
Mercury
Mercury
Formula:
Hg MW: 200,59 g/mol Boiling Pt: 356,5 °C (1013 hPa) Melting Pt: –39 °C Density: 13,54 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011035 CAS Number: 7439-97-6 EINECS: 231-106-7 UN: 2809 ADR: 8,III Merck Index: 13,05925 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...