Yttrium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.025 mm (0.001 in)
Supplier: Thermo Fisher Scientific
|
Warning
|
Packaged under argon.
12443.FF
12443.FI
12443.FFEA
198
EUR
12443.FF
12443.FI
Yttrium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.025 mm (0.001 in)
Yttrium
Purity: 99.9%
CAS No.: 7440-65-5
Physical Form: ≈0.07g/25x25mm, Packaged under argon
Odour: Odourless
UN No.: 3178
MDL No.: MFCD00011468
Formula:
Y Boiling Pt: 2927 °C (1013 hPa) Melting Pt: 1525 °C Density: 4,472 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011468 CAS Number: 7440-65-5 EINECS: 231-174-8 UN: 3178 ADR: 4.1,III Merck Index: 12,10240 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...