Tantalum ≥99.9% (metals basis), plate, Thickness: 6.35 mm (0.25 in)

Supplier: Thermo Fisher Scientific

14181.FF 14181.FI
14181.FFEA 692 EUR
14181.FF 14181.FI
Tantalum ≥99.9% (metals basis), plate, Thickness: 6.35 mm (0.25 in)
Tantalum

Purity: 99.9% CAS No.: 7440-25-7 Physical Form: ≈65.9g/25x25mm Odour: Odourless MDL No.: MFCD00011252

Formula: Ta
Boiling Pt: 5427 °C (1013 hPa)
Melting Pt: 2996 °C
Density: 16,69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR