Tantalum ≥99.9% (metals basis), wire, hard, Ø 0.025 mm (0.001 in)
Supplier: Thermo Fisher Scientific
40675.G1
40675.G6
40675.G1EA
272
EUR
40675.G1
40675.G6
Tantalum ≥99.9% (metals basis), wire, hard, Ø 0.025 mm (0.001 in)
Tantalum
Purity: 99.9%
CAS No.: 7440-25-7
Physical Form: ≈0.0082g/m, Temper: as drawn
Odour: Odourless
MDL No.: MFCD00011252
Microelectronics, contacts, surgical, high temperature
Formula:
Ta Boiling Pt: 5427 °C (1013 hPa) Melting Pt: 2996 °C Density: 16,69 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...