Iridium hard ≥99.8% (metals basis), foil, hard, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Scientific
11426.FF
11426.FI
11426.FFEA
1550
EUR
11426.FF
11426.FI
Iridium hard ≥99.8% (metals basis), foil, hard, Thickness: 0.127 mm (0.005 in)
Iridium
Purity: 99.8%
CAS No.: 7439-88-5
Physical Form: ≈1.8g/25x25mm, Temper: as rolled
MDL No.: MFCD00011062
Formula:
Ir MW: 192,22 g/mol Boiling Pt: 5300 °C (1013 hPa) Melting Pt: 2450 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011062 CAS Number: 7439-88-5 EINECS: 231-095-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...