7,8-Epoxyoctene ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1,2-Epoxy-7-octene

L07033.06 L07033.14
L07033.06EA 40.8 EUR
L07033.06 L07033.14
7,8-Epoxyoctene ≥97%
7,8-Epoxyoctene

Purity: 97% CAS No.: 19600-63-6 Molecular Formula: C8H14O Formula Weight: 126.20 Flash Point: 38°(100°F) Refractive Index: 1.4350 Boiling Point: 167° Beilstein Reference: 1305290 UN No.: 1993 MDL No.: MFCD00005156

Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 167 °C
Density: 0.850
Flash Pt: 38 °C (100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005156
CAS Number: 19600-63-6
EINECS: 243-178-7
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR