Chalcone 97%

Supplier: Thermo Scientific

Synonyms: 1,3-Diphenyl-2-propen-1-one, Phenyl Styryl ketone, 2-Benzalacetophenone, 2-Benzylideneacetophenone

159431000 159435000
ACRO159431000EA 61.3 EUR
ACRO159431000 ACRO159435000
Chalcone 97%
Chalcone
Formula: (C₆H₅)C(O)CHCH(C₆H₅)
MW: 208,26 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 53…58 °C
Density: 1,0712 g/cm³ (62.4 °C)
Flash Pt: >112 °C
MDL Number: MFCD00003082
CAS Number: 94-41-7
EINECS: 202-330-2
Merck Index: 13,02045

Order Now

Specification Test Results

Appearance Yellow Granular powder
Infrared spectrum Conforms
Melting point 53 to 58 °C
GC ≥96.0 %
Chloride (Cl) ≤0.01 %
Sulfate (SO4) ≤0.02 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR