Paraffin Oil heavy, liquid

Supplier: Roth Carl

8904.1 8904.2
ROTH8904.1EA 0 EUR
ROTH8904.1 ROTH8904.2
Paraffin Oil heavy, liquid
Paraffin Oil
Density: 0,86 g/cm³ (20 °C)
CAS Number: 8012-95-1
EINECS: 232-384-2
Merck Index: 13,07263

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR