Molecular sieve 3A (0.3 nm, 3 Å)
Supplier: Roth Carl
Synonyms:
Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate
P729.1
P729.2
ROTHP729.1EA
47.1
EUR
ROTHP729.1
ROTHP729.2
Molecular sieve 3A (0.3 nm, 3 Å)
Molecular sieve 3A (0.3 nm, 3 Å)
Formula:
KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O |
MDL Number:
MFCD00131613 CAS Number: 308080-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...