L(+)-Arabinose, Millipore®
Supplier: Merck Millipore Calbiochem
Synonyms:
Pectinose, Pectin Sugar
178680-100
178680-25
178680-100EA
508
EUR
178680-100
178680-25
L(+)-Arabinose, Millipore®
L(+)-Arabinose
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol Boiling Pt: 331 Melting Pt: 158…160 °C Density: 1.585 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00067709 CAS Number: 5328-37-0 EINECS: 226-214-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...