L(+)-Arabinose, Millipore®

Supplier: Merck


M-Clarity™ quality level = MQ100

1.01492.0100 1.01492.5000
1.01492.0100EA 408 EUR
1.01492.0100 1.01492.5000
L(+)-Arabinose, Millipore®
L(+)-Arabinose
Formula: C₅H₁₀O₅
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 158…160 °C
Density: 1,585 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00067709
CAS Number: 5328-37-0
EINECS: 226-214-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&vid=6b9738c515009e5720388ea307490836c3b8afa8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR