Starch, soluble
Supplier: SOUTHERN GROUP
Synonyms:
Starch
, Vitex® iodine indicator
, Amylodextrin
SGRO0519STEA
673
EUR
SGRO0519ST
Starch, soluble
Starch, soluble
Formula:
(C₆H₁₀O₅)n MW: 162,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00082026 CAS Number: 9005-84-9 EINECS: 232-686-4 Merck Index: 13,08878 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...