Canada balsam

Supplier: Roth Carl

8016.1 8016.2
ROTH8016.1EA 0 EUR
ROTH8016.1 ROTH8016.2
Canada balsam
Canada balsam
Density: 0,915 g/cm³ (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00132800
CAS Number: 8007-47-4
EINECS: 232-362-2
Merck Index: 13,00948

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR