Polyethylene glycol 1,500

Supplier: Roth Carl
Warning

0152.1 0152.2
ROTH0152.1EA 0 EUR
ROTH0152.1 ROTH0152.2
Polyethylene glycol 1,500
Polyethylene glycol 1,500
Formula: H(OCH₂CH₂)nOH
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 44…48 °C
Density: 1,13 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR