Polyethylene glycol 20,000
Supplier: Roth Carl
Synonyms:
PEG 20000, Ethylene glycol polymer 20000, PEG 20,000
0165.1
0165.2
ROTH0165.1EA
83.1
EUR
ROTH0165.1
ROTH0165.2
Polyethylene glycol 20,000
Polyethylene glycol 20,000
Formula:
H(OCH₂CH₂)nOH MW: 20000 g/mol Melting Pt: 58…63 °C Density: 1,13 g/cm³ (20 °C) Flash Pt: 260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...