Polyethylene glycol 4,000
Supplier: Roth Carl
Synonyms:
PEG 4,000
0156.1
0156.2
ROTH0156.1EA
0
EUR
ROTH0156.1
ROTH0156.2
Polyethylene glycol 4,000
Polyethylene glycol 4,000
Formula:
H(OCH₂CH₂)nOH MW: 4000 g/mol Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 53…58 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 250 Storage Temperature: Ambient |
MDL Number:
MFCD01779612 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...