Polyethylene glycol 8,000

Synonyms: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000

EMR037001 EMR037500
EURLEMR037001EA 0 EUR
EURLEMR037001 EURLEMR037500
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Boiling Pt: 250 °C (1013 hPa)
Density: 1,125 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779615
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 12,07729

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR