1-Iodopentane ≥98% stabilised

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Pentyl iodide

Stabilisation: Stabilised with copper

B20781.22 B20781.36
B20781.22EA 98.8 EUR
B20781.22 B20781.36
1-Iodopentane ≥98% stabilised
1-Iodopentane

Stabilized with copper


Purity: 98% CAS No.: 628-17-1 Molecular Formula: C5H11I Formula Weight: 198.05 Flash Point: 51°(123°F) Refractive Index: 1.4950 Odour: Characteristic Melting Point: -86° Boiling Point: 154-156° Beilstein Reference: 1730982 UN No.: 1993 MDL No.: MFCD00001100

Formula: C₅H₁₁I
Boiling Pt: 154…156 °C
Melting Pt: –86 °C
Density: 1,515
Flash Pt: 51 °C (123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001100
CAS Number: 628-17-1
EINECS: 211-030-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR