(±)-2-Mercaptopropionic acid 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-Mercaptopropionic acid
138791000
138795000
138790025
ACRO138791000EA
44.3
EUR
ACRO138791000
ACRO138795000
ACRO138790025
(±)-2-Mercaptopropionic acid 95%
(±)-2-Mercaptopropionic acid
Formula:
C₃H₆O₂S MW: 106.15 g/mol Boiling Pt: 102 °C (16 mmHg) Melting Pt: 10…14 °C Density: 1.19 g/cm³ Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004862 CAS Number: 79-42-5 EINECS: 201-206-5 UN: 2936 ADR: 6.1 II Merck Index: 12,09476 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4804 to 1.4824 (20°C, 589 nm) |
Titration Iodimetric | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...