DL-Malic acid, Millipore®
Supplier: Merck
|
Warning
|
Synonyms:
DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid
M-Clarity™ quality level = MQ100
1.00382.0250
1.00382.1000
1.00382.0250EA
66.3
EUR
1.00382.0250
1.00382.1000
DL-Malic acid, Millipore®
DL-Malic acid
Formula:
HOOCCH(OH)CH₂COOH MW: 134.09 g/mol Boiling Pt: 306 Melting Pt: 131…132 °C Density: 1.601 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064212 CAS Number: 617-48-1 EINECS: 230-022-8 Merck Index: 13,05730 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...