DL-Malic acid

Supplier: Roth Carl
Danger

Synonyms: DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid

3034.1 3034.2
ROTH3034.1EA 0 EUR
ROTH3034.1 ROTH3034.2
DL-Malic acid
DL-Malic acid
Formula: HOOCCH(OH)CH₂COOH
MW: 134.09 g/mol
Boiling Pt: 306
Melting Pt: 131…132 °C
Density: 1.601 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064212
CAS Number: 617-48-1
EINECS: 210-514-9
Merck Index: 13,05730
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR