(±)-Mercaptosuccinic acid for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: (±)-Mercapto-1,4-butanedioic acid, 2-Mercaptosuccinic acid, (±)-Thiomalic acid, DL-Mercaptosuccinic acid

M-Clarity™ quality level = MQ200

8.20763.0100 8.20763.5000 8.20763.9010
8.20763.0100EA 71.9 EUR
8.20763.0100 8.20763.5000 8.20763.9010
(±)-Mercaptosuccinic acid for synthesis, Sigma-Aldrich®
(±)-Mercaptosuccinic acid
Formula: C₄H₆O₄S
MW: 150.16 g/mol
Boiling Pt: 322
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00004860
CAS Number: 70-49-5
EINECS: 200-736-4
Merck Index: 12,09479

Order Now

Specification Test Results

Solubility 150 g/l (20 °C)
Melting Point 150 - 153 °C
Molar Mass 150.15 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR