Hydroquinone monomethyl ether 99%

Supplier: Thermo Scientific
Warning

Synonyms: p-Methoxyphenol, Mequinol, 4-Methoxyphenol, 1-Hydroxy-4-methoxybenzene, 4-Hydroxyanisole

126000010 126001000 126005000 126000051 126000250
ACRO126000010EA 164 EUR
ACRO126000010 ACRO126001000 ACRO126005000 ACRO126000051 ACRO126000250
Hydroquinone monomethyl ether 99%
Hydroquinone monomethyl ether
Formula: C₇H₈O₂
MW: 124,14 g/mol
Boiling Pt: 243 °C (760 mmHg)
Melting Pt: 56…57 °C
Density: 1,55 g/cm³ (20 °C)
Flash Pt: 133 °C
Storage Temperature: Ambient
MDL Number: MFCD00002332
CAS Number: 150-76-5
EINECS: 205-769-8

Order Now

Specification Test Results

Appearance White to light yellow Flakes
Infrared spectrum Conforms
Melting point 54 to 57 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR