γ-Terpinene 97% stabilised
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
gamma-Terpinene, p-Mentha-1,4-diene
207501000
207505000
ACRO207501000EA
60.5
EUR
ACRO207501000
ACRO207505000
γ-Terpinene 97% stabilised
γ-Terpinene
Formula:
C₁₀H₁₆ MW: 136,24 g/mol Boiling Pt: 182 °C (1013 hPa) Density: 0,847 g/cm³ Flash Pt: 51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001537 CAS Number: 99-85-4 EINECS: 202-794-6 UN: 2319 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to almost colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4720 to 1.4770 (20°C, 589 nm) |
Density | (20°C) 0.845 to 0.852 g/ml |
GC | ≥96.0 % |
Stabilizer | 0.01 to 0.1 % dl-alpha tocopherol |
Peroxide value | ≤30.0 meq/kg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...