1,3,5,7-Cyclooctatetraene 98% stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Cyclooctatetraene, COT
160570025
160570100
160572500
ACRO160570025EA
111
EUR
ACRO160570025
ACRO160570100
ACRO160572500
1,3,5,7-Cyclooctatetraene 98% stabilised
1,3,5,7-Cyclooctatetraene
Formula:
C₈H₈ MW: 104.15 g/mol Boiling Pt: 142…143 °C (760 mmHg) Melting Pt: –27 °C Density: 0.92 g/cm³ Flash Pt: 22 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004161 CAS Number: 629-20-9 EINECS: 211-080-3 UN: 2358 ADR: 3 II |
Specification Test Results
Appearance | Turbid yellow to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5365 to 1.5385 (20°C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | 0.1 % hydroquinone |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...