Dicyclopentadiene 95% stabilised

Supplier: Thermo Fisher Scientific
Danger


Stabilisation: Stabilised with 4-tert-butylpyrocatechol (TBC) 100-200 ppm

150760010 150760025 150761000 150760100
ACRO150760010EA 106 EUR
ACRO150760010 ACRO150760025 ACRO150761000 ACRO150760100
Dicyclopentadiene 95% stabilised
Dicyclopentadiene
Formula: C₁₀H₁₂
Boiling Pt: 170 °C (760 mmHg)
Melting Pt: 11…13 °C
Density: 0,982 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00078246
CAS Number: 77-73-6
EINECS: 201-052-9
UN: 2048
ADR: 3,III
Merck Index: 13,02767

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5120 (20°C, 589 nm)
Water ≤0.03 % (K.F.)
Density (20°C) 0.975 to 0.989 kg/l
GC ≥91 % (endo-Dicyclopentadiene)
Color scale ≤50 APHA
Stabilizer 100 to 200 ppm p-tert-Butylcatechol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR